Anti-MYD88 Mouse Monoclonal Antibody [clone: OTI2B2]

Supplier: OriGene
ORIGTA502117EA 581 EUR
ORIGTA502117
Anti-MYD88 Mouse Monoclonal Antibody [clone: OTI2B2]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: MYD88 (myeloid differentiation primary response 88)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR