Anti-SNX8 Mouse Monoclonal Antibody [clone: OTI4B4]

Supplier: OriGene
ORIGTA502134EA 581 EUR
ORIGTA502134
Anti-SNX8 Mouse Monoclonal Antibody [clone: OTI4B4]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: SNX8 (sorting nexin 8)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR