Anti-GMDS Mouse Monoclonal Antibody [clone: OTI2A1]

Supplier: OriGene
TA502141
ORIGTA502141EA 621 EUR
ORIGTA502141
Anti-GMDS Mouse Monoclonal Antibody [clone: OTI2A1]
Antibodies
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: GMDS (GDP-mannose 4,6-dehydratase)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR