Anti-GALE Mouse Monoclonal Antibody [clone: OTI1C4]

Supplier: OriGene
ORIGTA502146EA 586 EUR
ORIGTA502146
Anti-GALE Mouse Monoclonal Antibody [clone: OTI1C4]
Antibodies
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150

Type: Primary
Antigen: GALE (UDP-galactose-4-epimerase)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR