Anti-HDAC6 Mouse Monoclonal Antibody [clone: OTI3F9]

Supplier: OriGene
ORIGTA502222EA 603 EUR
ORIGTA502222
Anti-HDAC6 Mouse Monoclonal Antibody [clone: OTI3F9]
Antibodies
  • Recommended Dilutions: Western Blot: 1:500; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: HDAC6 (histone deacetylase 6)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR