Anti-CTDSP1 Mouse Monoclonal Antibody [clone: OTI1G2]

Supplier: OriGene
ORIGTA502227EA 603 EUR
ORIGTA502227
Anti-CTDSP1 Mouse Monoclonal Antibody [clone: OTI1G2]
Antibodies
Recommended Dilutions: Western Blot: 1:500; Flow Cytometry: 1:100

Type: Primary
Antigen: CTDSP1 (CTD (carboxy-terminal domain, RNA polymerase II, polypeptide A) small phosphatase 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR