Anti-DNAJB1 Mouse Monoclonal Antibody [clone: OTI1B12]

Supplier: OriGene
ORIGTA502245EA 603 EUR
ORIGTA502245
Anti-DNAJB1 Mouse Monoclonal Antibody [clone: OTI1B12]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: DNAJB1 (DnaJ (Hsp40) homolog, subfamily B, member 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR