Anti-MMAB Mouse Monoclonal Antibody [clone: OTI1G3]

Supplier: OriGene
TA502265
ORIGTA502265EA 616 EUR
ORIGTA502265
Anti-MMAB Mouse Monoclonal Antibody [clone: OTI1G3]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: MMAB (methylmalonic aciduria (cobalamin deficiency) cblB type)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR