Anti-CTDSP1 Mouse Monoclonal Antibody [clone: OTI1F4]

Supplier: OriGene
TA502289
ORIGTA502289EA 616 EUR
ORIGTA502289
Anti-CTDSP1 Mouse Monoclonal Antibody [clone: OTI1F4]
Antibodies
Recommended Dilutions: Western Blot: 1:500

Type: Primary
Antigen: CTDSP1 (CTD (carboxy-terminal domain, RNA polymerase II, polypeptide A) small phosphatase 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR