Anti-BCHE Mouse Monoclonal Antibody [clone: OTI1E1]

Supplier: OriGene
ORIGTA502307EA 608 EUR
ORIGTA502307
Anti-BCHE Mouse Monoclonal Antibody [clone: OTI1E1]
Antibodies
  • Recommended Dilutions: Western Blot: 1:500-2000; Flow Cytometry: 1:100

Type: Primary
Antigen: BCHE (butyrylcholinesterase)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR