Anti-NUDT6 Mouse Monoclonal Antibody [clone: OTI10F12]

Supplier: OriGene
ORIGTA502335EA 603 EUR
ORIGTA502335
Anti-NUDT6 Mouse Monoclonal Antibody [clone: OTI10F12]
Antibodies
Recommended Dilutions: Western Blot: 1:500

Type: Primary
Antigen: NUDT6 (nudix (nucleoside diphosphate linked moiety X)-type motif 6)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR