Anti-VCAM1 Mouse Monoclonal Antibody [clone: OTI4E8]

Supplier: OriGene
ORIGTA502360EA 603 EUR
ORIGTA502360
Anti-VCAM1 Mouse Monoclonal Antibody [clone: OTI4E8]
Antibodies
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150

Type: Primary
Antigen: VCAM1 (vascular cell adhesion molecule 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR