Anti-PIP4K2A Mouse Monoclonal Antibody [clone: OTI3A9]

Supplier: OriGene
ORIGTA502369EA 608 EUR
ORIGTA502369
Anti-PIP4K2A Mouse Monoclonal Antibody [clone: OTI3A9]
Antibodies
Recommended Dilutions: Western Blot: 1:200-500; Flow Cytometry: 1:100

Type: Primary
Antigen: PIP4K2A (phosphatidylinositol-5-phosphate 4-kinase, type II, alpha)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR