Anti-MSI1 Mouse Monoclonal Antibody [clone: OTI3G4]

Supplier: OriGene
ORIGTA502370EA 603 EUR
ORIGTA502370
Anti-MSI1 Mouse Monoclonal Antibody [clone: OTI3G4]
Antibodies
  • Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: MSI1 (musashi RNA-binding protein 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR