Anti-PRKCE Mouse Monoclonal Antibody [clone: OTI1B7]

Supplier: OriGene
ORIGTA502395EA 603 EUR
ORIGTA502395
Anti-PRKCE Mouse Monoclonal Antibody [clone: OTI1B7]
Antibodies
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: PRKCE (protein kinase C, epsilon)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR