Anti-RASSF5 Mouse Monoclonal Antibody [clone: OTI1C6]

Supplier: OriGene
ORIGTA502434EA 603 EUR
ORIGTA502434
Anti-RASSF5 Mouse Monoclonal Antibody [clone: OTI1C6]
Antibodies
Recommended Dilutions: Western Blot: 1:500

Type: Primary
Antigen: RASSF5 (Ras association (RalGDS/AF-6) domain family member 5)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR