Anti-PNMA3 Mouse Monoclonal Antibody [clone: OTI2E6]

Supplier: OriGene
ORIGTA502458EA 603 EUR
ORIGTA502458
Anti-PNMA3 Mouse Monoclonal Antibody [clone: OTI2E6]
Antibodies
Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:150

Type: Primary
Antigen: PNMA3 (paraneoplastic Ma antigen 3)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR