Anti-NDUFB9 Mouse Monoclonal Antibody [clone: OTI3E11]

Supplier: OriGene
ORIGTA502503EA 603 EUR
ORIGTA502503
Anti-NDUFB9 Mouse Monoclonal Antibody [clone: OTI3E11]
Antibodies
Recommended Dilutions: Western Blot: 1:200-500

Type: Primary
Antigen: NDUFB9 (NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 9, 22kDa)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR