Anti-MCTS1 Mouse Monoclonal Antibody [clone: OTI5B5]

Supplier: OriGene
ORIGTA502506EA 603 EUR
ORIGTA502506
Anti-MCTS1 Mouse Monoclonal Antibody [clone: OTI5B5]
Antibodies
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:150

Type: Primary
Antigen: MCTS1 (malignant T cell amplified sequence 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR