Anti-HPGD Mouse Monoclonal Antibody [clone: OTI2C10]

Supplier: OriGene
ORIGTA502561EA 608 EUR
ORIGTA502561
Anti-HPGD Mouse Monoclonal Antibody [clone: OTI2C10]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: HPGD (hydroxyprostaglandin dehydrogenase 15-(NAD))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR