Anti-HOXC11 Mouse Monoclonal Antibody [clone: OTI2H9]

Supplier: OriGene
ORIGTA502575EA 586 EUR
ORIGTA502575
Anti-HOXC11 Mouse Monoclonal Antibody [clone: OTI2H9]
Antibodies
Recommended Dilutions: Western Blot: 1:200-500; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: HOXC11 (homeobox C11)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR