Anti-LACTB2 Mouse Monoclonal Antibody [clone: OTI4E7]

Supplier: OriGene
ORIGTA502583EA 581 EUR
ORIGTA502583
Anti-LACTB2 Mouse Monoclonal Antibody [clone: OTI4E7]
Antibodies
  • Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: LACTB2 (lactamase, beta 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR