Anti-ARFGAP1 Mouse Monoclonal Antibody [clone: OTI1H4]

Supplier: OriGene
ORIGTA502594EA 603 EUR
ORIGTA502594
Anti-ARFGAP1 Mouse Monoclonal Antibody [clone: OTI1H4]
Antibodies
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: ARFGAP1 (ADP-ribosylation factor GTPase activating protein 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR