Anti-PON1 Mouse Monoclonal Antibody [clone: OTI1F7]

Supplier: OriGene
ORIGTA502611EA 603 EUR
ORIGTA502611
Anti-PON1 Mouse Monoclonal Antibody [clone: OTI1F7]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: PON1 (paraoxonase 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR