Anti-DSTN Mouse Monoclonal Antibody [clone: OTI1C5]

Supplier: OriGene
TA502661
ORIGTA502661EA 616 EUR
ORIGTA502661
Anti-DSTN Mouse Monoclonal Antibody [clone: OTI1C5]
Antibodies
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: DSTN (destrin (actin depolymerizing factor))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR