Anti-TBCEL Mouse Monoclonal Antibody [clone: OTI2A9]

Supplier: OriGene
ORIGTA502672EA 603 EUR
ORIGTA502672
Anti-TBCEL Mouse Monoclonal Antibody [clone: OTI2A9]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: TBCEL (tubulin folding cofactor E-like)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR