Anti-MGRN1 Mouse Monoclonal Antibody [clone: OTI1E12]

Supplier: OriGene
ORIGTA502685EA 581 EUR
ORIGTA502685
Anti-MGRN1 Mouse Monoclonal Antibody [clone: OTI1E12]
Antibodies
  • Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: MGRN1 (mahogunin ring finger 1, E3 ubiquitin protein ligase)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR