Anti-PDLIM2 Mouse Monoclonal Antibody [clone: OTI3B5]

Supplier: OriGene
ORIGTA502710EA 603 EUR
ORIGTA502710
Anti-PDLIM2 Mouse Monoclonal Antibody [clone: OTI3B5]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: PDLIM2 (PDZ and LIM domain 2 (mystique))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR