Anti-STAT5A Mouse Monoclonal Antibody [clone: OTI3D2]

Supplier: OriGene
TA502741
ORIGTA502741EA 616 EUR
ORIGTA502741
Anti-STAT5A Mouse Monoclonal Antibody [clone: OTI3D2]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: STAT5A (signal transducer and activator of transcription 5A)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR