Anti-LMCD1 Mouse Monoclonal Antibody [clone: OTI2G5]

Supplier: OriGene
ORIGTA502790EA 603 EUR
ORIGTA502790
Anti-LMCD1 Mouse Monoclonal Antibody [clone: OTI2G5]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: LMCD1 (LIM and cysteine-rich domains 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR