Anti-FGFR2 Mouse Monoclonal Antibody [clone: OTI1D3]

Supplier: OriGene
ORIGTA502797EA 603 EUR
ORIGTA502797
Anti-FGFR2 Mouse Monoclonal Antibody [clone: OTI1D3]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: FGFR2 (fibroblast growth factor receptor 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR