Anti-ALDH1A3 Mouse Monoclonal Antibody [clone: OTI4B6]

Supplier: OriGene
ORIGTA502805EA 608 EUR
ORIGTA502805
Anti-ALDH1A3 Mouse Monoclonal Antibody [clone: OTI4B6]
Antibodies
Recommended Dilutions: Western Blot: 1:200; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: ALDH1A3 (aldehyde dehydrogenase 1 family, member A3)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR