Anti-PNMT Mouse Monoclonal Antibody [clone: OTI1D2]

Supplier: OriGene
ORIGTA502822EA 608 EUR
ORIGTA502822
Anti-PNMT Mouse Monoclonal Antibody [clone: OTI1D2]
Antibodies
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: PNMT (phenylethanolamine N-methyltransferase)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR