Anti-HDHD2 Mouse Monoclonal Antibody [clone: OTI4G3]

Supplier: OriGene
TA502834
ORIGTA502834EA 616 EUR
ORIGTA502834
Anti-HDHD2 Mouse Monoclonal Antibody [clone: OTI4G3]
Antibodies
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: HDHD2 (haloacid dehalogenase-like hydrolase domain containing 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR