Anti-PLEKHA3 Mouse Monoclonal Antibody [clone: OTI6E4]

Supplier: OriGene
ORIGTA502876EA 581 EUR
ORIGTA502876
Anti-PLEKHA3 Mouse Monoclonal Antibody [clone: OTI6E4]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: PLEKHA3 (pleckstrin homology domain containing, family A (phosphoinositide binding specific) member 3)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR