Anti-FGF21 Mouse Monoclonal Antibody [clone: OTI2B9]

Supplier: OriGene
ORIGTA502897EA 581 EUR
ORIGTA502897
Anti-FGF21 Mouse Monoclonal Antibody [clone: OTI2B9]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: FGF21 (fibroblast growth factor 21)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR