Anti-MGLL Mouse Monoclonal Antibody [clone: OTI2C9]

Supplier: OriGene
ORIGTA502931EA 603 EUR
ORIGTA502931
Anti-MGLL Mouse Monoclonal Antibody [clone: OTI2C9]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: MGLL (monoglyceride lipase)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR