Anti-LGR5 Mouse Monoclonal Antibody [clone: OTI11B5]

Supplier: OriGene
ORIGTA502947EA 603 EUR
ORIGTA502947
Anti-LGR5 Mouse Monoclonal Antibody [clone: OTI11B5]
Antibodies
Recommended Dilutions: Western Blot: 1:500; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: LGR5 (leucine-rich repeat containing G protein-coupled receptor 5)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR