Anti-HDHD2 Mouse Monoclonal Antibody [clone: OTI2E9]

Supplier: OriGene
ORIGTA503014EA 603 EUR
ORIGTA503014
Anti-HDHD2 Mouse Monoclonal Antibody [clone: OTI2E9]
Antibodies
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: HDHD2 (haloacid dehalogenase-like hydrolase domain containing 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR