Anti-AP2M1 Mouse Monoclonal Antibody [clone: OTI1C8]

Supplier: OriGene
TA503018
ORIGTA503018EA 616 EUR
ORIGTA503018
Anti-AP2M1 Mouse Monoclonal Antibody [clone: OTI1C8]
Antibodies
Recommended Dilutions: Western Blot: 1:500-2000; Immunofluorescence: 1:100

Type: Primary
Antigen: AP2M1 (adaptor-related protein complex 2, mu 1 subunit)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR