Anti-AP2M1 Mouse Monoclonal Antibody [clone: OTI1E9]

Supplier: OriGene
ORIGTA503019EA 608 EUR
ORIGTA503019
Anti-AP2M1 Mouse Monoclonal Antibody [clone: OTI1E9]
Antibodies
Recommended Dilutions: Western Blot: 1:200-500; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: AP2M1 (adaptor-related protein complex 2, mu 1 subunit)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR