Anti-GSPT2 Mouse Monoclonal Antibody [clone: OTI2G9]

Supplier: OriGene
ORIGTA503066EA 608 EUR
ORIGTA503066
Anti-GSPT2 Mouse Monoclonal Antibody [clone: OTI2G9]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: GSPT2 (G1 to S phase transition 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR