Anti-LENG1 Mouse Monoclonal Antibody [clone: OTI1E6]

Supplier: OriGene
ORIGTA503084EA 581 EUR
ORIGTA503084
Anti-LENG1 Mouse Monoclonal Antibody [clone: OTI1E6]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: LENG1 (leukocyte receptor cluster (LRC) member 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR