Anti-TOMM34 Mouse Monoclonal Antibody [clone: OTI2A9]

Supplier: OriGene
TA503090
ORIGTA503090EA 621 EUR
ORIGTA503090
Anti-TOMM34 Mouse Monoclonal Antibody [clone: OTI2A9]
Antibodies
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Immunofluorescence: 1:50-100; Flow Cytometry: 1:100

Type: Primary
Antigen: TOMM34 (translocase of outer mitochondrial membrane 34)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR