Anti-ILVBL Mouse Monoclonal Antibody [clone: OTI3E9]

Supplier: OriGene
ORIGTA503141EA 603 EUR
ORIGTA503141
Anti-ILVBL Mouse Monoclonal Antibody [clone: OTI3E9]
Antibodies
  • Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:500

Type: Primary
Antigen: ILVBL (ilvB (bacterial acetolactate synthase)-like)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR