Anti-PSMD2 Mouse Monoclonal Antibody [clone: OTI1B5]

Supplier: OriGene
TA503220
ORIGTA503220EA 616 EUR
ORIGTA503220
Anti-PSMD2 Mouse Monoclonal Antibody [clone: OTI1B5]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100

Type: Primary
Antigen: PSMD2 (proteasome (prosome, macropain) 26S subunit, non-ATPase, 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR