Anti-SIRT5 Mouse Monoclonal Antibody [clone: OTI2E12]

Supplier: OriGene
ORIGTA503292EA 581 EUR
ORIGTA503292
Anti-SIRT5 Mouse Monoclonal Antibody [clone: OTI2E12]
Antibodies
Recommended Dilutions: Western Blot: 1:500; Flow Cytometry: 1:100

Type: Primary
Antigen: SIRT5 (sirtuin 5)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR