Anti-DOK2 Mouse Monoclonal Antibody [clone: OTI2A3]

Supplier: OriGene
ORIGTA503302EA 603 EUR
ORIGTA503302
Anti-DOK2 Mouse Monoclonal Antibody [clone: OTI2A3]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: DOK2 (docking protein 2, 56kDa)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR