Anti-XPNPEP3 Mouse Monoclonal Antibody [clone: OTI5G2]

Supplier: OriGene
ORIGTA503304EA 603 EUR
ORIGTA503304
Anti-XPNPEP3 Mouse Monoclonal Antibody [clone: OTI5G2]
Antibodies
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: XPNPEP3 (X-prolyl aminopeptidase (aminopeptidase P) 3, putative)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR