Anti-COASY Mouse Monoclonal Antibody [clone: OTI2H1]

Supplier: OriGene
ORIGTA503343EA 603 EUR
ORIGTA503343
Anti-COASY Mouse Monoclonal Antibody [clone: OTI2H1]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: COASY (CoA synthase)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR